Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADNT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203721.1 | NP_001190650.1 | 6 - 7 | 948 | 94 | |
NM_001340300.1 | NP_001328407.1 | 6 - 7 | 1281 | 94 | ||
NM_116340.4 | NP_192019.1 | 6 - 7 | 895 | 94 | ||
GenBank mRNA | AF360168.1 | - | 6 - 7 | 890 | 94 | |
AF412085.1 | - | 6 - 7 | 891 | 94 | ||
AM931440.1 | - | 6 - 7 | 636 | 94 | ||
AY056343.1 | - | 6 - 7 | 636 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |