Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116342.3 | NP_192021.1 | 11 - 12 | 1001 | 79 | |
GenBank mRNA | AY062667.1 | - | 11 - 12 | 959 | 79 | |
AY093316.1 | - | 10 - 11 | 818 | 79 | ||
X63895.1 | - | 903 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |