Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116342.3 | NP_192021.1 | 5 - 6 | 475 | 69 | |
GenBank mRNA | AY062667.1 | - | 5 - 6 | 433 | 69 | |
AY093316.1 | - | 4 - 5 | 292 | 69 | ||
X63895.1 | - | 377 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |