Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340321.1 | NP_001329345.1 | 8 - 9 | 707 | 75 | |
NM_116378.4 | NP_192057.1 | 8 - 9 | 738 | 75 | ||
GenBank mRNA | AY054516.1 | - | 8 - 9 | 689 | 75 | |
AY081667.1 | - | 7 - 8 | 604 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |