Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116386.4 | NP_567213.1 | 2 - 3 | 439 | 83 | |
GenBank mRNA | AY050341.1 | - | 2 - 3 | 371 | 83 | |
AY086463.1 | - | 2 - 3 | 392 | 83 | ||
BT020609.1 | - | 2 - 3 | 327 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |