Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116386.4 | NP_567213.1 | 3 - 4 | 527 | 74 | |
GenBank mRNA | AY050341.1 | - | 3 - 4 | 459 | 74 | |
AY086463.1 | - | 3 - 4 | 480 | 74 | ||
BT020609.1 | - | 3 - 4 | 415 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |