Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116402.4 | NP_192081.1 | 4 - 5 | 857 | 80 | |
GenBank mRNA | AF480165.1 | - | 4 - 5 | 712 | 80 | |
AK229522.1 | - | 4 - 5 | 740 | 80 | ||
BT008704.1 | - | 4 - 5 | 740 | 80 | ||
BT029024.1 | - | 4 - 5 | 712 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |