Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340338.1 | NP_001328373.1 | 6 - 7 | 1700 | 60 | |
NM_116412.2 | NP_192091.1 | 6 - 7 | 1711 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |