Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340339.1 | NP_001329375.1 | 2 - 3 | 1070 | 137 | |
NM_116413.2 | NP_192092.1 | 6 - 7 | 1714 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |