Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SWN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340355.1 | NP_001328437.1 | 5 - 6 | 760 | 69 | |
NM_001340356.1 | NP_001328436.1 | 5 - 6 | 757 | 69 | ||
NM_001340357.1 | NP_001328435.1 | 5 - 6 | 757 | 69 | ||
NM_116433.3 | NP_567221.1 | 5 - 6 | 777 | 69 | ||
GenBank mRNA | AF100163.1 | - | 5 - 6 | 731 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |