Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203726.1 | NP_001190655.1 | 14 - 15 | 1823 | 68 | |
NM_116437.3 | NP_192115.1 | 12 - 13 | 1773 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |