Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203726.1 | NP_001190655.1 | 9 - 10 | 934 | 132 | |
NM_116437.3 | NP_192115.1 | 7 - 8 | 884 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |