Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203727.2 | NP_001190656.1 | 9 - 10 | 2360 | 103 | |
NM_116438.2 | NP_192116.1 | 9 - 10 | 2370 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |