Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116439.3 | NP_567222.1 | 6 - 7 | 658 | 139 | |
GenBank mRNA | AF130849.1 | - | 6 - 7 | 625 | 139 | |
DQ086841.1 | - | 6 - 7 | 563 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |