Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116461.4 | NP_192137.1 | 7 - 8 | 1906 | 79 | |
GenBank mRNA | AY051001.1 | - | 7 - 8 | 1680 | 79 | |
AY056784.1 | - | 7 - 8 | 1705 | 79 | ||
AY142511.1 | - | 7 - 8 | 1466 | 79 | ||
KJ138769.1 | - | 7 - 8 | 1466 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |