Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340373.1 | NP_001329308.1 | 3 - 4 | 649 | 152 | |
NM_001340374.1 | NP_001329307.1 | 2 - 3 | 716 | 152 | ||
NM_116472.4 | NP_192148.2 | 3 - 4 | 823 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |