Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036498.3 | NP_001031575.1 | 20 - 21 | 2350 | 66 | |
NM_001036499.1 | NP_001031576.1 | 20 - 21 | 2286 | 66 | ||
NM_001203732.1 | NP_001190661.1 | 20 - 21 | 2348 | 66 | ||
NM_116491.3 | NP_567243.1 | 19 - 20 | 2444 | 66 | ||
GenBank mRNA | AJ318017.1 | - | 18 - 19 | 2145 | 66 | |
AK317123.1 | - | 20 - 21 | 2287 | 66 | ||
AY046030.1 | - | 19 - 20 | 2367 | 66 | ||
AY133878.1 | - | 18 - 19 | 2111 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |