Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNE12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036500.2 | NP_001031577.1 | 2 - 3 | 669 | 74 | |
NM_001125447.1 | NP_001118919.1 | 3 - 4 | 427 | 74 | ||
NM_116493.3 | NP_567245.1 | 2 - 3 | 718 | 74 | ||
GenBank mRNA | AF367328.1 | - | 2 - 3 | 599 | 74 | |
AK316748.1 | - | 2 - 3 | 670 | 74 | ||
AY143951.1 | - | 2 - 3 | 484 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |