Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036501.2 | NP_001031578.1 | 10 - 11 | 1343 | 74 | |
NM_116494.5 | NP_192169.1 | 9 - 10 | 1427 | 74 | ||
GenBank mRNA | AY072135.1 | - | 9 - 10 | 1274 | 74 | |
AY113992.1 | - | 9 - 10 | 1174 | 74 | ||
Z95352.1 | - | 1266 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |