Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULTR3;2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116504.3 | NP_192179.1 | 7 - 8 | 1203 | 103 | |
GenBank mRNA | AB004060.1 | - | 7 - 8 | 1163 | 103 | |
AK229084.1 | - | 7 - 8 | 1203 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |