Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116532.3 | NP_192207.1 | 3 - 4 | 577 | 91 | |
GenBank mRNA | AF389278.1 | - | 3 - 4 | 427 | 91 | |
AY093754.1 | - | 3 - 4 | 418 | 91 | ||
S40550.1 | - | 3 - 4 | 427 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |