Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AOP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340416.1 | NP_001319854.1 | 4 - 5 | 1217 | 80 | |
NM_001340417.1 | NP_001319855.1 | 4 - 5 | 1221 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |