Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XTH9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036504.1 | NP_001031581.1 | 2 - 3 | 340 | 97 | |
NM_116559.3 | NP_192230.1 | 2 - 3 | 672 | 97 | ||
GenBank mRNA | AB493675.1 | - | 2 - 3 | 270 | 97 | |
AY044333.1 | - | 2 - 3 | 270 | 97 | ||
AY072353.1 | - | 2 - 3 | 337 | 97 | ||
BT002199.1 | - | 2 - 3 | 270 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |