Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tic20-IV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116570.5 | NP_192241.1 | 2 - 3 | 252 | 81 | |
GenBank mRNA | AF361633.1 | - | 2 - 3 | 301 | 81 | |
AY056069.1 | - | 2 - 3 | 190 | 81 | ||
AY084543.1 | - | 2 - 3 | 317 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |