Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116594.5 | NP_567258.1 | 13 - 14 | 1682 | 99 | |
GenBank mRNA | AF360372.1 | - | 13 - 14 | 1358 | 99 | |
AK316725.1 | - | 13 - 14 | 1399 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |