Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116659.5 | NP_567262.1 | 1 - 2 | 687 | 125 | |
GenBank mRNA | AY085333.1 | - | 1 - 2 | 192 | 125 | |
AY091099.1 | - | 1 - 2 | 177 | 125 | ||
AY122960.1 | - | 1 - 2 | 151 | 125 | ||
AY572784.1 | - | 1 - 2 | 151 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |