Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPK22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340496.1 | NP_001319866.1 | 3 - 4 | 906 | 97 | |
NM_001340497.1 | NP_001329214.1 | 3 - 4 | 969 | 97 | ||
NM_001340498.1 | NP_001329213.1 | 4 - 5 | 825 | 97 | ||
NM_001340499.1 | NP_001329212.1 | 3 - 4 | 906 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |