Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203747.1 | NP_001190676.1 | 4 - 5 | 891 | 94 | |
NM_001340519.1 | NP_001329057.1 | 4 - 5 | 937 | 94 | ||
NM_116730.5 | NP_192401.5 | 4 - 5 | 604 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |