Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340521.1 | NP_001328262.1 | 4 - 5 | 402 | 98 | |
NM_116734.3 | NP_567276.1 | 5 - 6 | 559 | 98 | ||
GenBank mRNA | AY045901.1 | - | 5 - 6 | 487 | 98 | |
AY113948.1 | - | 5 - 6 | 466 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |