Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203749.1 | NP_001190678.1 | 2 - 3 | 489 | 101 | |
NM_116741.4 | NP_567283.1 | 2 - 3 | 683 | 101 | ||
GenBank mRNA | AY039856.1 | - | 2 - 3 | 415 | 101 | |
BT002241.1 | - | 2 - 3 | 358 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |