Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203749.1 | NP_001190678.1 | 7 - 8 | 1549 | 87 | |
NM_116741.4 | NP_567283.1 | 7 - 8 | 1743 | 87 | ||
GenBank mRNA | AY039856.1 | - | 7 - 8 | 1475 | 87 | |
BT002241.1 | - | 7 - 8 | 1418 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |