Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340542.1 | NP_001329202.1 | 8 - 9 | 1250 | 96 | |
NM_116758.4 | NP_192428.2 | 9 - 10 | 1547 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |