Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACS11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116873.2 | NP_567330.1 | 1 - 2 | 285 | 93 | |
GenBank mRNA | AF332405.1 | - | 1 - 2 | 135 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |