Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116904.6 | NP_192575.3 | 9 - 10 | 1833 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |