Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORTHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084888.1 | NP_001078357.1 | 1 - 2 | 398 | 145 | |
NM_001340597.1 | NP_001328494.1 | 1 - 2 | 475 | 145 | ||
NM_001340598.1 | NP_001328497.1 | 1 - 2 | 459 | 145 | ||
NM_001340599.1 | NP_001328496.1 | 1 - 2 | 438 | 145 | ||
NM_001340600.1 | NP_001328495.1 | 1 - 2 | 438 | 145 | ||
NM_116928.4 | NP_192599.2 | 1 - 2 | 475 | 145 | ||
GenBank mRNA | AK175637.1 | - | 1 - 2 | 399 | 145 | |
BT022101.1 | - | 1 - 2 | 398 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |