Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APRL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116962.4 | NP_192632.2 | 1 - 2 | 503 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |