Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116971.6 | NP_192641.2 | 5 - 6 | 652 | 103 | |
GenBank mRNA | AK227049.1 | - | 5 - 6 | 489 | 103 | |
AY091058.1 | - | 5 - 6 | 485 | 103 | ||
AY133739.1 | - | 5 - 6 | 462 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |