Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRZ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104097.6 | NP_175628.2 | 10 - 11 | 2401 | 83 | |
GenBank mRNA | AY070061.1 | - | 10 - 11 | 2366 | 83 | |
AY117303.1 | - | 10 - 11 | 2346 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |