Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRZ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104097.6 | NP_175628.2 | 1 - 2 | 364 | 77 | |
GenBank mRNA | AY070061.1 | - | 1 - 2 | 329 | 77 | |
AY117303.1 | - | 1 - 2 | 309 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |