Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRZ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104097.6 | NP_175628.2 | 8 - 9 | 1379 | 117 | |
GenBank mRNA | AY070061.1 | - | 8 - 9 | 1344 | 117 | |
AY117303.1 | - | 8 - 9 | 1324 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |