Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333566.1 | NP_001319211.1 | 2 - 3 | 256 | 120 | |
GenBank mRNA | DQ459170.1 | - | 2 - 3 | 256 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |