Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPK18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104229.4 | NP_175756.2 | 8 - 9 | 1478 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |