Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333607.1 | NP_001321367.1 | 10 - 11 | 1030 | 67 | |
NM_104250.5 | NP_175776.2 | 10 - 11 | 1037 | 67 | ||
GenBank mRNA | AY568526.1 | - | 9 - 10 | 911 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |