Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333607.1 | NP_001321367.1 | 8 - 9 | 933 | 119 | |
NM_104250.5 | NP_175776.2 | 8 - 9 | 940 | 119 | ||
GenBank mRNA | AY568526.1 | - | 7 - 8 | 814 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |