Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH7B4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104287.5 | NP_175812.1 | 12 - 13 | 1458 | 98 | |
NM_179476.3 | NP_849807.1 | 11 - 12 | 1344 | 98 | ||
GenBank mRNA | AF378873.1 | - | 12 - 13 | 1316 | 98 | |
AJ584645.1 | - | 11 - 12 | 1226 | 98 | ||
AK230363.1 | - | 3123 | 98 | |||
AY048242.1 | - | 11 - 12 | 1341 | 98 | ||
AY091032.1 | - | 11 - 12 | 1343 | 98 | ||
AY102145.1 | - | 11 - 12 | 1224 | 98 | ||
AY117345.1 | - | 11 - 12 | 1224 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |