Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH7B4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104287.5 | NP_175812.1 | 13 - 14 | 1560 | 82 | |
NM_179476.3 | NP_849807.1 | 12 - 13 | 1446 | 82 | ||
GenBank mRNA | AF378873.1 | - | 13 - 14 | 1418 | 82 | |
AJ584645.1 | - | 12 - 13 | 1328 | 82 | ||
AK230363.1 | - | 3225 | 82 | |||
AY048242.1 | - | 12 - 13 | 1443 | 82 | ||
AY091032.1 | - | 12 - 13 | 1445 | 82 | ||
AY102145.1 | - | 12 - 13 | 1326 | 82 | ||
AY117345.1 | - | 12 - 13 | 1326 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |