Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH7B4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104287.5 | NP_175812.1 | 6 - 7 | 680 | 67 | |
NM_179476.3 | NP_849807.1 | 5 - 6 | 566 | 67 | ||
GenBank mRNA | AF378873.1 | - | 6 - 7 | 538 | 67 | |
AJ584645.1 | - | 5 - 6 | 448 | 67 | ||
AK230363.1 | - | 2345 | 67 | |||
AY048242.1 | - | 5 - 6 | 563 | 67 | ||
AY091032.1 | - | 5 - 6 | 565 | 67 | ||
AY102145.1 | - | 5 - 6 | 446 | 67 | ||
AY117345.1 | - | 5 - 6 | 446 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |