Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333633.1 | NP_001320241.1 | 1 - 2 | 72 | 109 | |
NM_104299.3 | NP_175823.1 | 1 - 2 | 86 | 109 | ||
GenBank mRNA | AB073184.1 | - | 1 - 2 | 72 | 109 | |
AF492758.1 | - | 1 - 2 | 43 | 109 | ||
BT024654.1 | - | 1 - 2 | 39 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |