Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333641.1 | NP_001321996.1 | 12 - 13 | 1270 | 84 | |
NM_104312.3 | NP_175836.1 | 13 - 14 | 1232 | 84 | ||
GenBank mRNA | AF316501.1 | - | 13 - 14 | 1058 | 84 | |
BT025983.1 | - | 13 - 14 | 1038 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |