Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333641.1 | NP_001321996.1 | 6 - 7 | 495 | 82 | |
NM_104312.3 | NP_175836.1 | 6 - 7 | 562 | 82 | ||
GenBank mRNA | AF316501.1 | - | 6 - 7 | 388 | 82 | |
BT025983.1 | - | 6 - 7 | 368 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |